ContactUsLinkComponent
Female Threaded Tee Coupling, 3/8" NPT(F), PTFE
Female Threaded Tee Coupling, 3/8" NPT(F), PTFE
目录编号 MFLB34030-21
供应商:  MASTERFLEX PREMIUM
undefined
Female Threaded Tee Coupling, 3/8" NPT(F), PTFE
目录编号 MFLB34030-21
供应商:  MASTERFLEX PREMIUM
关于此项目
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings are designed for more permanent installations. Choose the material and size that meets your application needs.
  • Connection Type:
    Threaded Union
  • Flow pattern:
    T
  • Thread size NPT(F) (port 1):
    ⅜"
  • Thread size NPT(F) (port 2):
    ⅜"
  • 说明:
    Female Threaded Tee Coupling, ⅜" NPT(F), PTFE
  • 顺序:
    18
  • Environmentally Preferable:
  • 型号:
    34030-21
  • 灭菌:
    Autoclave or ethylene oxide (ETO)
  • 包装规格:
    1
  • Fitting material:
    PTFE
  • Thread size NPT(F) (port 3):
    ⅜"