ContactUsLinkComponent
McMillan Flow Power supply/output cable (0 to 5 VDC), 115 VAC.
McMillan Flow Power supply/output cable (0 to 5 VDC), 115 VAC.
Catalog # MFLB32707-50
Supplier:  MASTERFLEX PREMIUM
undefined
McMillan Flow Power supply/output cable (0 to 5 VDC), 115 VAC.
Catalog # MFLB32707-50
Supplier:  MASTERFLEX PREMIUM
About this item
Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.
  • Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
  • 说明:
    Power supply/output cable (0 to 5 VDC), 115 VAC.
  • Environmentally Preferable:
  • 质保:
    1 year
  • 包装规格:
    1