Masterflex® 带倒钩的 NPT 母螺纹直型转接头,Avantor®
05, Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/4" NPT(F); 100/PK, 1/4", Thread to Barb, Nylon, Straight, 1/4"
目录编号 MFLB31701-85
供应商: MASTERFLEX PREMIUM
Jump to:
Product Details & Documents
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
规格
- Barb size/tubing int.Ø (port 2):1/4"
- Connection Type:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(F) (port 1):1/4"
- 说明:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/4" NPT(F); 100/PK
- 顺序:05
- Fitting material:Nylon
- Cat. No.:MFLB31701-85
- 灭菌:Ethylene oxide (ETO) only
- 质保:1 year
- 型号:31701-85