黄铜压缩NPT(M)转接器
Compression pipe adapter, Brass, 1/4"×1/8", Each, Compression to Thread, Brass, Straight, 1/8"
Catalog # MFLB31412-69
Supplier: MASTERFLEX PREMIUM
Jump to:
Product Details & Documents
Create a secure connection with compression fittings.
- Constructed of durable brass
良好的耐化学性。在77ºF ( 25ºC)时可达到最大值400 psi。可用温度范围为 -85 至482ºF (-65 至 250ºC).
Specifications
- Connection Type:Compression to Thread
- Flow pattern:Straight
- Thread size NPT(M) (port 2):1/8"
- 说明:Compression pipe adapter, Brass, 1/4"×1/8", Each
- Fitting material:Brass
- Cat. No.:MFLB31412-69
- 质保:LTD life time