ContactUsLinkComponent
Masterflex® Pump Extension Cable, RJ-12, 15-ft (4.5 m) Length; 1/Ea
Masterflex® Pump Extension Cable, RJ-12, 15-ft (4.5 m) Length; 1/Ea
目录编号 MFLA77095-04
undefined
Masterflex® Pump Extension Cable, RJ-12, 15-ft (4.5 m) Length; 1/Ea
目录编号 MFLA77095-04
关于此项目
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
  • Choose accessories and replacement parts suitable for your Masterflex® pump
  • Increase flexibility with extended cable lengths
Convenient accessories to make your job easier.
  • 说明:
    Optional extension cable, RJ12; 15 ft (4,5 m).
  • Environmentally Preferable:
  • 包装规格:
    1