Masterflex® Fitting, Polypropylene, Straight, Female Luer to Low-Profile Hosebarb, 1/16"; 25/PK
Catalog # MFLA45508-12
Supplier: MASTERFLEX PROPRIETARY
CE certificate
About this item
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
- Barb size/tubing int.Ø (port 1):¹⁄₁₆"
- Connection Type:Female Luer to Barb
- Flow pattern:Straight
- 说明:Fitting, Polypropylene, Straight, Female Luer to Low-Profile Hosebarb, ¹⁄₁₆"; 25/PK
- 顺序:12
- Environmentally Preferable:
- 型号:45508-12
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- 包装规格:25
- Fitting material:Polypropylene