Masterflex® Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/8" ID x 1/2" NPT(M)
MFLA34030-82
CE certificate
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/8" ID x 1/2" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Barb size/tubing int.Ø (port 2):3/8"
- Connection Type:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/2"
- 说明:Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/8" ID x 1/2" NPT(M)
- 顺序:10
- Environmentally Preferable:
- No. of ports:2
- 灭菌:Autoclave or ethylene oxide (ETO)
- 质保:2 years
- 包装规格:1
- Fitting material:PTFE