Masterflex® Fitting, Black PVDF, One-Way, Stopcock, Male Luer Lock to Female Luer Lock; 10/PK
目录编号 MFLA31200-91
关于此项目
Build leakproof fluid handling systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
旋塞有天然PVDF塞体和聚丙烯承插件,或全部黑色PVDF构造。天然PVDF旋塞有双向和三向配置;黑色PVDF旋塞是单向。
- Flow pattern:One-Way
- 说明:Luer stopcock, 1-way, male luer lock x female luer, black PVDF, pack of 10
- 顺序:03
- Environmentally Preferable:
- 型号:31200-91
- 浸湿材料:Black PVDF
- 包装规格:10
- Inlet port:Female Luer
- Outlet port:Male Luer Lock