ContactUsLinkComponent
Masterflex® Fitting, Nylon, Straight, Male Threaded Adapter, 1/2" NPT(M); 10/PK
Masterflex® Fitting, Nylon, Straight, Male Threaded Adapter, 1/2" NPT(M); 10/PK
目录编号 MFLA30629-02
undefined
Masterflex® Fitting, Nylon, Straight, Male Threaded Adapter, 1/2" NPT(M); 10/PK
目录编号 MFLA30629-02
关于此项目
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/2" NPT(M)
  • Constructed of durable nylon
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ½"
  • Thread size NPT(M) (port 2):
    ½"
  • 说明:
    Fitting, Nylon, Straight, Male Threaded Adapter, ½" NPT(M); 10/PK
  • 顺序:
    19
  • Environmentally Preferable:
  • 型号:
    30629-02
  • 灭菌:
    Ethylene oxide (ETO) only
  • 包装规格:
    10
  • Fitting material:
    Nylon