Masterflex® 倒刺异径直通接头,Avantor®
01, Barbed fittings, reducing connector, stainless steel, tubing ID: ½×¼", ½", ¼", Barbed reducer, Stainless steel 316, Straight
Jump to:
Product Details & Documents
Ideal for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilise via ethylene oxide, autoclave, or gamma irradiation.
Documents
Certificates
按批号搜索规格
- Barb size/tubing int.Ø (port 1):½"
- Barb size/tubing int.Ø (port 2):¼"
- Connection Type:Barbed reducer
- Flow pattern:Straight
- 说明:Barbed fittings, reducing connector, stainless steel, tubing ID: ½×¼"
- 顺序:01
- Fitting material:Stainless steel 316
- Cat. No.:MFLA30620-11
- 灭菌:Autoclave, ethylene oxide (ETO), or gamma
- 质保:—
- 型号:30620-11
- Orifice dimensions:4,763 mm (0,1875")