Masterflex® 聚碳酸酯歧管,Avantor®
Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK, Female Luer, Female Luer, 5 Ports, 360 Degree Rotation
Catalog # MFLA30600-43
Supplier: MASTERFLEX PROPRIETARY
Jump to:
Product Details & Documents
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
聚碳酸酯歧管带有聚氨酯管茎,高密度聚乙烯把手。端口有母luer锁。单独订购歧管或10件一箱的方便装。
Specifications
- Flow pattern:5 Ports, 360 Degree Rotation
- 说明:Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
- Inlet port:Female Luer
- Outlet port:Female Luer
- Cat. No.:MFLA30600-43
- 质保:Out of box
- 浸湿材料:Polycarbonate