Masterflex® Footswitch, Momentary Start/Stop Operation, DB25; 6 ft
Catalog # MFLA07523-92
Supplier: MASTERFLEX PROPRIETARY
About this item
Easily control your Masterflex® drive with compatible footswitch.
- Choose footswitch with the correct connection for your Masterflex® pump
Add convenience to repetitive dispensing with a foot-operated on/off switch. Keep hands free to efficiently move containers during routine fill applications or to keep containers stable during the filling process.
- 控制类型:Analog Variable Speed
- 说明:Footswitch, momentary start/stop, DB25, 6' cable
- Environmentally Preferable:
- 型号:07523-92
- 质保:2 years
- 包装规格:1