Orders LinkContactUsLinkComponent
Masterflex® 电刷盖和电刷,Avantor®
Masterflex® 电刷盖和电刷,Avantor®
Replacement brushes for L/S drive series: 07554-80, -85, -90, -95
目录编号 MFLA07520-06
Masterflex® 电刷盖和电刷,Avantor®
目录编号 MFLA07520-06
Masterflex® 电刷盖和电刷,Avantor®
Masterflex® 电刷盖和电刷,Avantor®
Replacement brushes for L/S drive series: 07554-80, -85, -90, -95
Jump to:
Price
询价
Log in 查看库存
1 * 2 件
Product Details & Documents
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
规格
  • 说明:
    Replacement brushes for L/S drive series: 07554-80, -85, -90, -95