Masterflex® 电刷盖和电刷,Avantor®
Drive Replacement Brush Cap, Series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300
MFLA07520-03
Quickly and easily replace components of your Masterflex system.
- Choose replacement brushes and caps based on your Masterflex L/S drive
- Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
- 说明:Drive Replacement Brush Cap
- 兼容:Series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300
- Cat. No.:MFLA07520-03
- 质保:1 year