Masterflex® R系列无阀活塞泵头,Avantor®
02, Series R Pump Head Adapter Kit; Series R Head to Series Q Drives
MFLA07117-99
Compatible with Masterflex® L/S® or series Q drive.
- 该泵头与Masterflex® L/S®或Q系列驱动器兼容;
- 无阀,不存在堵塞或磨损问题;
- 可用于低流量输送;
- Kynar®泵体可抵挡强腐蚀性化学剂;
- R系列泵头装配有可防止堵塞的旋转活塞,非常适用于泵送粘度达500厘泊的流体。
压力接头可接1/4英寸外径管。为完善成套装配,需订购R系列泵头和一台Masterflex(R) L/S或Q系列驱动器。使用转换接头配件连接泵头和驱动器。
不同的液体可能有不同的流量特性。
- 说明:Series R Pump Head Adapter Kit; Series R Head to Series Q Drives
- 顺序:02
- Cat. No.:MFLA07117-99
- Inlet connection:Series R Head to Series Q Drives Adapter
- 型号:07117-99