Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 24
Catalog # MFLA07024-81
Supplier: MASTERFLEX PROPRIETARY
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- 说明:Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 24 Tubing
- 顺序:30
- Environmentally Preferable:
- 型号:07024-81
- Open-head sensor:No
- Tubing sizes accepted:L/S 24
- 质保:2 years
- 包装规格:1
- Housing material:PC
- Rotor material:Cold-rolled steel (CRS)