Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Short Shaft, SS Rotor
Catalog # MFLA07019-85
Supplier: MASTERFLEX PROPRIETARY
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Constructed of durable stainless steel (SS)
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- 说明:Standard Pump Head, Replacement Rotor Assembly, Short Shaft, SS Rotor; 1/Ea
- 顺序:28
- Environmentally Preferable:
- 型号:07019-85
- Open-head sensor:No
- Tubing sizes accepted:I/P 73, 82
- 质保:2 years
- 包装规格:1
- Rotor material:Stainless Steel