Masterflex® Fitting, Polypropylene, Tee, Hose Barb Union, 1/16" ID, 25/PK
Catalog # MFLA06365-77
Supplier: MASTERFLEX PROPRIETARY
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Barb size/tubing int.Ø (port 1):¹⁄₁₆"
- Barb size/tubing int.Ø (port 2):¹⁄₁₆"
- Connection Type:Barbed Union
- Flow pattern:T
- 说明:Fitting, Polypropylene, Tee, Hose Barb Union, ¹⁄₁₆" ID, 25/PK
- 顺序:08
- Environmentally Preferable:
- 型号:06365-77
- Orifice dimensions:0,03125"
- 灭菌:Autoclave or ethylene oxide (ETO)
- 包装规格:25
- Barb size/tubing int.Ø (port 3):¹⁄₁₆"
- Fitting material:Polypropylene