Masterflex® 倒刺交叉接头,Avantor®
19, Fitting, Polypropylene, Cross Connector, Hosebarb, ½" ID, Cleanroom Packed; 20/Bg, ½", ½", ½", ¹⁄₁₆", Barbed Union, Polypropylene, X
Supplier: MASTERFLEX PROPRIETARY
Jump to:
Product Details & Documents
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. Cross connectors feature four hose barb ports.
Documents
Certificates
按批号搜索规格
- Barb size/tubing int.Ø (port 1):½"
- Barb size/tubing int.Ø (port 2):½"
- Connection Type:Barbed Union
- Flow pattern:X
- 说明:Fitting, Polypropylene, Cross Connector, Hosebarb, ½" ID, Cleanroom Packed; 20/Bg
- 顺序:19
- Barb size/tubing int.Ø (port 3):½"
- Barb size/tubing int.Ø (port 4):¹⁄₁₆"
- Fitting material:Polypropylene
- Cat. No.:MFLA40726-74
- 质保:Out of box
- 型号:40726-74