Masterflex® 直角倒刺接头,Avantor®
20, Fitting, Polypropylene, Elbow, Hose Barb Union, ³⁄₁₆" ID; 10/PK, ³⁄₁₆", ³⁄₁₆", Barbed Union, Polypropylene, Elbow
Supplier: MASTERFLEX PROPRIETARY
Jump to:
Product Details & Documents
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
Documents
Certificates
Search by Lot NumberSpecifications
- Barb size/tubing int.Ø (port 1):³⁄₁₆"
- Barb size/tubing int.Ø (port 2):³⁄₁₆"
- Connection Type:Barbed Union
- Flow pattern:Elbow
- 说明:Fitting, Polypropylene, Elbow, Hose Barb Union, ³⁄₁₆" ID; 10/PK
- 顺序:20
- Fitting material:Polypropylene
- Cat. No.:MFLA40622-75
- 灭菌:Autoclave or ethylene oxide (ETO)
- 型号:40622-75