Volumetric flasks, class B
06, 500 ml, 19/26, 0,50 ± ml
Catalog # VITL675941
Supplier: VITLAB
Jump to:
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
- 模拟器:19/26
- 容量:500 ml
- 顺序:06
- 误差:0,50 ± ml