Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
05, 250 ml, 19/26, 0,30 ± ml
目录编号 VITL674941
供应商:   VITLAB
Volumetric flasks, class B
目录编号 VITL674941
供应商:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
05, 250 ml, 19/26, 0,30 ± ml
Jump to:
Price
询价
Log in 查看库存
1 * 2 件
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
规格
  • 模拟器:
    19/26
  • 容量:
    250 ml
  • 顺序:
    05
  • 误差:
    0,30 ± ml