Orders LinkContactUsLinkComponent
Volumetric flasks, class B
04, 100 ml, 14/23, 0,20 ± ml
Catalog # VITL673941
Supplier:   VITLAB
Volumetric flasks, class B
Catalog # VITL673941
Supplier:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
04, 100 ml, 14/23, 0,20 ± ml
Price
Inquire For Price
Log in for availability
1 * 2 件
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
  • 模拟器:
    14/23
  • 容量:
    100 ml
  • 顺序:
    04
  • 误差:
    0,20 ± ml