Volumetric flasks, class B
03, 50 ml, 10/19, 0,12 ± ml
目录编号 VITL672941
供应商: VITLAB
Jump to:
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
规格
- 模拟器:10/19
- 容量:50 ml
- 顺序:03
- 误差:0,12 ± ml