Volumetric flasks, class B
02, 25 ml, 10/19, 0,08 ± ml
VITL671941
: VITLAB
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
- 模拟器:10/19
- 容量:25 ml
- 顺序:02
- 误差:0,08 ± ml