Thread adaptors, TYPE A, b.safe
06, PP, GL 45 (male), 38×3 (female)
目录编号 BOHLM615-29
供应商: BOHLENDER
Jump to:
Product Details & Documents
All adaptors of this type are supplied with a male thread, a female thread and recessed grips.
- Stay flexible and independent
- Recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
规格
- 螺纹:GL 45 (male), 38×3 (female)
- 材质:PP
- 顺序:06