Thread adaptors, TYPE B, b.safe
04, PP, S 60/61 (male), Tri-Sure 2" (male)
Catalog # BOHLM615-12
Supplier: BOHLENDER
Jump to:
Product Details & Documents
All adaptors of this type are supplied with two male threads and recessed grips.
- Stay flexible and independent
- With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
Specifications
- 螺纹:S 60/61 (male), Tri-Sure 2" (male)
- 材质:PP
- 顺序:04