Thread adaptors, TYPE D, b.safe
01, PTFE/PPS, GL 45 (female), NS 29/32
目录编号 BOHLM520-01
供应商: BOHLENDER
Product Details & Documents
For assembly of components with ground joint cone NS 29/32 on bottles and vessels with GL 45 thread.
- Stay flexible and independent
- With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
规格
- 插座:NS 29/32
- 螺纹:GL 45 (female)
- 说明:PTFE/PPS
- 顺序:01
Accessories
产品信息
- 目录编号说明可用库存Price