L-组氨酸 USP, Multi-Compendial, J.T.Baker®
4130, 6,78 kg, 6,78 kg
供应商: Avantor
Jump to:
Product Details & Documents
同义词: (S)-2-氨基-3-(4-咪唑基)丙酸
- CAS号:71-00-1
- 化学式:C₆H₉N₃O₂
- MDL 编号:MFCD00064315
- Molecular Weight:155.16 g/mol
- 默克索引:13,04741
- 熔点:272…273 °C
- 密度:1,44 g/cm³ (20 °C)
- :Management of Change (MOC) category = R
- 储存温度:Ambient
- EINECS:200-745-3
Documents
规格
规格测试结果
- USP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):12.6 - 14.0 Degree
- USP - pH:7.0 - 8.5
- USP - Loss on Drying at 105°C:≤ 0.2 %
- USP - Residue on Ignition:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):11.4 - 12.4 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Assay (C6H9N3O2) (dried basis):99.0 - 101.0 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Chloride (Cl):≤ 0.021 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Identification:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP- Optical Rotation [Å]²º^D (+):11.8 - 12.8 Degree
- JP - Related Substances:Passes Test
- JP - pH:7.0 - 8.5
- JP - Residue on Ignition:≤ 0.10 %
- JP - Sulfate (SO₄):≤ 0.028 %
- Bacterial endotoxins, EU/g:≤ 10
规格
- 包装规格:6,78 kg
- 顺序:4130
- Pk Info:6,78 kg
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



