China
Orders LinkContactUsLinkComponent
DEC-HAND®洗手液
DEC-HAND®洗手液
04, DEC-HAND® 946 毫升,用于 ASEPTI-CLEANSE 系统的瓶装,无菌
目录编号 VELTDH-10-E
供应商:   VELTEK ASSOCIATES
DEC-HAND®洗手液
目录编号 VELTDH-10-E
供应商:  VELTEK ASSOCIATES
DEC-HAND®洗手液
DEC-HAND®洗手液
04, DEC-HAND® 946 毫升,用于 ASEPTI-CLEANSE 系统的瓶装,无菌
Jump to:
Price
¥16,042.00
Log in 查看库存
1 * 12 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DEC-HAND®是一种酒精洗手液,它符合制药,生物技术,医疗设备和健康护理领域等领域相关标准。DEC-HAND®在所有设施中都有多种用途,并且符合运输到分级区域的要求。
  • 可作为即用洗手液
  • ABCD洁净室介入系统内采用四层包装
  • 以ISO 5(A/B级, 即此前的100级)对其进行灌装操作
  • 已过滤
  • 进行γ射线辐照,确保其无菌水平达到10-6
  • 可确保无菌性和保存期限
473Ml装产品可配连接式泵头或者直接安装在墙面分配器上。此产品可以完全可追溯并且对无菌性和保存期限进行验证。产品交付时配备批次产品专属相关证书
规格
  • 说明:
    DEC-HAND® 946 毫升,用于 ASEPTI-CLEANSE 系统的瓶装,无菌
  • 顺序:
    04
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.