Orders LinkContactUsLinkComponent
洁净室专用纸,PUREImage
洁净室专用纸,PUREImage
23, Latex-free paper, three hole punched, 白色, 80 g/m²
Catalog # VWRU87003-430
   Sustainable Packaging
洁净室专用纸,PUREImage
Catalog # VWRU87003-430
   Sustainable Packaging
洁净室专用纸,PUREImage
洁净室专用纸,PUREImage
23, Latex-free paper, three hole punched, 白色, 80 g/m²
Price
¥5,208.00
Log in for availability
1 * 2.500 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
洁净室专用纸适用于防化学和颗粒污染都非常重要的洁净室环境。
  • 高拉伸强度和卓越的耐热性能,能够实现更佳复制和打印效率
  • PE结合科技,实现低颗粒水平,不挥发残留物及离子提取物
  • 独特的精确切割技术确保持续施胶和清洁度
  • 纸张在裁切前会进行双面纸幅清洁
在ISO 4级(FED-STD-209E 10级/M2,5)洁净室生产和清洁,并在ISO 5级(FED-STD-209E 100级)洁净室进行双层袋包装。纸张适用于ISO 4级 (FED-STD-209E 10级)到ISO 6级 (FED-STD-209E 1000级)洁净室。
Poly paper excels in printability. Synthetic paper excels in tensile strength.
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
Packaging: 包装:纸张包装格式为250张/包,10包/箱。
Specifications
  • 重量:
    80 g/m²
  • 颜色:
    白色
  • 说明:
    Latex-free paper, three hole punched
  • 顺序:
    23
  • Cat. No.:
    VWRU87003-430
  • Environmentally Preferable:
    Sustainable Packaging