VWR® Basic, Overalls, Dual-Zip Closure
01, 白色, M
目录编号 VWRI113-9010
供应商: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
规格
- 衣服尺寸:M
- 颜色:白色
- 顺序:01
- 重量:40 g/m²
- Cat. No.:VWRI113-9010
- 关闭类型:配有拉链封口
- Garment application:General Purpose|Laboratory|Industrial