防水温度计,Traceable®
05, Customer-Specified 3 to 4 Points Calibration Per Themometer
目录编号 VWRU10171-578
Product Details & Documents
该款防水防震的温度计是所有危险水银仪器的安全替代产品。其探头可耐受大多数化学品腐蚀。
- 防水结构,适用于湿实验室、冲洗区和所有野外应用
- 能够调取最大/最小温度记忆
- 读数每秒一次
不锈钢探头(长×直径:203×3.5 mm)
Delivery information: Supplied with probe and cover, silver-oxide battery and Traceable® certificate of calibration.
规格
- 说明:Customer-Specified 3 to 4 Points Calibration Per Themometer
- 顺序:05
Accessories
产品信息
- 目录编号说明可用库存Price