VWR®, Sample library tubes
02, 8-well tubes strips, 透明
Catalog # VWRU83009-680
Supplier: VWR BRAND PRIVATE LABEL USA
Jump to:
Product Details & Documents
PP, clear.
- Autoclavable and freezable (–80 °C)
- Printed with an alpha numeric grid that helps to find a sample easily
These 1,2 ml sample library tubes are supplied in a 96 tube format for storage of specimens. The rack is designed to meet SBS standards and can be used in robotic applications.
Delivery information: Supplied in 8×12 racks with clear, snug fit lid.
Specifications
- 颜色:透明
- 说明:8-well tubes strips
- 顺序:02
Accessories
Product Information
- Catalog #顺序说明AvailabilityPrice