Orders LinkContactUsLinkComponent
洁净室专用纸,PUREImage
23, Latex-free paper, A5 size, synthetic substrate, 蓝色, 105 g/m²
目录编号 VWRI115-1953
   Sustainable Packaging
供应商:   VWR Collection
洁净室专用纸,PUREImage
目录编号 VWRI115-1953
供应商:  VWR Collection
   Sustainable Packaging
洁净室专用纸,PUREImage
洁净室专用纸,PUREImage
23, Latex-free paper, A5 size, synthetic substrate, 蓝色, 105 g/m²
Jump to:
Price
¥4,991.00
Log in 查看库存
1 * 5.000 件
Product Details & Documents
洁净室专用纸适用于防化学和颗粒污染都非常重要的洁净室环境。
  • 高拉伸强度和卓越的耐热性能,能够实现更佳复制和打印效率
  • PE结合科技,实现低颗粒水平,不挥发残留物及离子提取物
  • 独特的精确切割技术确保持续施胶和清洁度
  • 纸张在裁切前会进行双面纸幅清洁
在ISO 4级(FED-STD-209E 10级/M2,5)洁净室生产和清洁,并在ISO 5级(FED-STD-209E 100级)洁净室进行双层袋包装。纸张适用于ISO 4级 (FED-STD-209E 10级)到ISO 6级 (FED-STD-209E 1000级)洁净室。
Poly paper excels in printability. Synthetic paper excels in tensile strength.
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
 : 包装:纸张包装格式为250张/包,10包/箱。
规格
  • 重量:
    105 g/m²
  • 颜色:
    蓝色
  • 说明:
    Latex-free paper, A5 size, synthetic substrate
  • 顺序:
    23
  • Cat. No.:
    VWRI115-1953
  • Environmentally Preferable:
    Sustainable Packaging