Accessories
10, Discharge tube for Dispensette® S Trace Analysis, PT/IR valve spring, without recirculation valve, 105 mm length, 105 mm
目录编号 BRND708022
供应商: BRAND
Jump to:
Product Details & Documents
规格
- 说明:Discharge tube for Dispensette® S Trace Analysis, PT/IR valve spring, without recirculation valve, 105 mm length
- 长度:105 mm
- 顺序:10