Accessories
07, Discharge tube, without recirculation valve, shape: standard tip, 105 mm length, for Dispensette S Organic 5 and 10 ml
目录编号 BRND708014
供应商: BRAND
Jump to:
Product Details & Documents
规格
- 说明:Discharge tube, without recirculation valve, shape: standard tip, 105 mm length, for Dispensette S Organic 5 and 10 ml
- 顺序:07