Accessories
06, Discharge tube, without recirculation valve, shape: standard tip, 135 mm length, for Dispensette S 25.50 and 100 ml
目录编号 BRND708008
供应商: BRAND
Jump to:
Product Details & Documents
规格
- 说明:Discharge tube, without recirculation valve, shape: standard tip, 135 mm length, for Dispensette S 25.50 and 100 ml
- 顺序:06