VWR® Surface Sampling Sponge Kits
09, Surface sampling sponges pre-moistened with Dey/Engley neutralising buffer
目录编号 VWRC300-0231
供应商: VWR Chemicals
Jump to:
Product Details & Documents
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
- Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
- Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
- Larger surface in comparison with normal sampling swabs
This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.
Certifications: HACCP, USDA, ACIA and CFIA requirements.
Caution: Gloves should be worn during all sampling procedures.
Documents
Certificates
按批号搜索Basic information about microbial monitoring of cleanrooms The Use of Contact Slides for Microbial Monitoring of Surface and Liquids in Food Industry
CoC VWR Humidified Surface Sampling Sponge with Neutralising Buffer Sampling Bag CoC VWR Humidified Surface Sampling Sponge with Neutralising Sterile Buffer Sampling Bag CoC VWR Surface Sampling Sponges CoC_VWRC300-0230_J095776 CoC_VWRC300-0230_J096983
More
Neutralizing Buffer Composition
Additional Product Details
规格
- 说明:Surface sampling sponges pre-moistened with Dey/Engley neutralising buffer
- 顺序:09