Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
02, Conductivity standard, 84 µS/cm, 10
目录编号 METT30111140
供应商:   METTLER TOLEDO
Conductivity standards
目录编号 METT30111140
供应商:  METTLER TOLEDO
CAS号:  
Conductivity standards
Conductivity standards
02, Conductivity standard, 84 µS/cm, 10
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 10 件
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
规格
规格
  • 包装规格:
    10
  • 说明:
    Conductivity standard
  • 顺序:
    02
  • 电导率:
    84 µS/cm
规格