Capillary columns, HP-PLOT Al₂O₃ S
13, with two particle traps, Capillary, HP-PLOT Al₂O₃ S, 15,00 µm, 0,53 mm, 50 m
HEWL19095P-S25PT
These columns are with middle range polarity. It is ideal for hydrocarbon analysis, and for resolving acetylene from butane and propylene from isobutane.
Aluminium oxide deactivated with sodium sulfate
- 列格式:Capillary
- 薄膜厚度:15,00 µm
- 内径:0,53 mm
- 说明:with two particle traps
- 长度:50 m
- 顺序:13
- Phase:HP-PLOT Al₂O₃ S