Orders LinkContactUsLinkComponent
洁净室实验服,VWR® Maximum,SF
14, 白色, 5XL
  VWRU414004-703
洁净室实验服,VWR® Maximum,SF
  VWRU414004-703
洁净室实验服,VWR® Maximum,SF
洁净室实验服,VWR® Maximum,SF
14, 白色, 5XL
¥1,955.00
1 * 25 件
VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness, and comfort properties.
  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level I Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
在经ISO认证的工厂进行测试和制造,并有着严格的程序控制。由独立实验室测试进行批量控制和检验。
  • 衣服尺寸:
    5XL
  • 颜色:
    白色
  • 顺序:
    14
  • Cat. No.:
    VWRU414004-703
  • 关闭类型:
    带按扣紧固
  • Garment application:
    Cleanroom|Laboratory
  • 袋:
    带口袋