China
Orders LinkContactUsLinkComponent
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
05, COD reagents, UHR, 250 - 15,000 mg/l, Reactor digestion, 1
目录编号 HACHTNT82306
供应商:   HACH
Test kits, COD, TNT plus®
目录编号 HACHTNT82306
供应商:  HACH
CAS号:  
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
05, COD reagents, UHR, 250 - 15,000 mg/l, Reactor digestion, 1
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary
规格
规格
  • 化学方法:
    Reactor digestion
  • 包装规格:
    1
  • 说明:
    COD reagents, UHR
  • 顺序:
    05
  • 范围:
    250 - 15,000 mg/l
规格