Test kits, COD, TNT plus®
05, COD reagents, UHR, 250 - 15,000 mg/l, Reactor digestion, 1
目录编号 HACHTNT82306
供应商: HACH
Jump to:
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are colour coded by parameter and range
- No reagent blank necessary
规格
规格
- 化学方法:Reactor digestion
- 包装规格:1
- 说明:COD reagents, UHR
- 顺序:05
- 范围:250 - 15,000 mg/l