China
Orders LinkContactUsLinkComponent
Test strips
03, Total hardness test strips, individually wrapped, 1000 tests, 0 - 425 mg/L, TS, 1.000
  HACH2793828
 :   HACH
Test strips
  HACH2793828
 :  HACH
 :  
Test strips
Test strips
03, Total hardness test strips, individually wrapped, 1000 tests, 0 - 425 mg/L, TS, 1.000
 :  
1 * 1.000 件
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
  • 包装规格:
    1.000
  • 说明:
    Total hardness test strips, individually wrapped, 1000 tests
  • 顺序:
    03
  • 类型:
    TS
  • 范围:
    0 - 425 mg/L