03, Total hardness test strips, individually wrapped, 1000 tests, 0 - 425 mg/L, TS, 1.000
:
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
Portable - ideal for field testing of surface water
Easy to use
包装规格:
1.000
说明:
Total hardness test strips, individually wrapped, 1000 tests