D-(+)-木糖 ≥98.0% (by HPLC)
3645, Glass bottle, 500 g
目录编号 TCIAX0019-500G
供应商: TCI
Jump to:
Product Details & Documents
- CAS号:58-86-6
- 化学式:C₅H₁₀O₅
- MDL 编号:MFCD00151475
- Molecular Weight:150.13 g/mol
- 默克索引:14,10087
- 沸点:331 °C (1013 hPa)
- 熔点:154 °C
- 密度:1,525 g/cm³ (20 °C)
- 储存温度:Ambient
- EINECS:200-400-7
Documents
规格
规格测试结果
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:146.0 to 153.0 deg-C
- Specific rotation [a]20/D:+19.0 to +21.0 deg (C=10, H2O)
- Solubility in Water:almost transparency
规格
- 包装类型:Glass bottle
- 包装规格:500 g
- 顺序:3645
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



