Orders LinkContactUsLinkComponent
刻度移液管,型号3,B级
08, 50 ml, 0,2 ml, 0,3 ± ml
Catalog # VWRI612-4128
Supplier:   VWR Collection
刻度移液管,型号3,B级
Catalog # VWRI612-4128
Supplier:  VWR Collection
刻度移液管,型号3,B级
刻度移液管,型号3,B级
08, 50 ml, 0,2 ml, 0,3 ± ml
Price
¥968.00
Log in for availability
1 * 2 件
Product Details & Documents
Borosilicate glass 9.1.
  • 校准后交付 (TD, Ex)
  • 琥珀色刻度标识
  • 棉塞末端(产品不包括棉塞)
DIN 12696
 : DIN 12696
Specifications
  • 容量:
    50 ml
  • 顺序:
    08
  • 分度:
    0,2 ml
  • 误差:
    0,3 ± ml