刻度移液管,型号3,B级
06, 20 ml, 0,1 ml, 0,15 ± ml
目录编号 VWRI612-4126
供应商: VWR Collection
Jump to:
Product Details & Documents
Borosilicate glass 9.1.
- 校准后交付 (TD, Ex)
- 琥珀色刻度标识
- 棉塞末端(产品不包括棉塞)
DIN 12696
: DIN 12696
规格
- 容量:20 ml
- 顺序:06
- 分度:0,1 ml
- 误差:0,15 ± ml
Borosilicate glass 9.1.
DIN 12696
: DIN 12696