Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
移液管和移液器
刻度移液管
刻度移液管,型号3,B级
刻度移液管,型号3,B级
刻度移液管,型号3,B级
05, 10 ml, 0,1 ml, 0,075 ± ml
VWRI612-4125
:
VWR Collection
刻度移液管,型号3,B级
VWRI612-4125
:
VWR Collection
Expand image
刻度移液管,型号3,B级
05, 10 ml, 0,1 ml, 0,075 ± ml
Add to cart
¥263.00
Unit of Measure
1 * 3 件
-
+
Add to cart
Borosilicate glass 9.1.
校准后交付 (TD, Ex)
琥珀色刻度标识
棉塞末端(产品不包括棉塞)
DIN 12696
Certifications:
DIN 12696
Documents
CoC Graduated Pipettes Type 3 Class B Borosilicate 9.1 glass
coc_VWRI612-4122_GL0625
coc_VWRI612-4122_GL4525
coc_VWRI612-4123_GL1919
coc_VWRI612-4123_GL3125
Product Literature
VWR® for Liquid Handling
VWR® Glassware Solutions
容量:
10 ml
顺序:
05
分度:
0,1 ml
误差:
0,075 ± ml