刻度移液管,型号3,B级
04, 5 ml, 0,1 ml, 0,045 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate glass 9.1.
- 校准后交付 (TD, Ex)
- 琥珀色刻度标识
- 棉塞末端(产品不包括棉塞)
DIN 12696
Certifications: DIN 12696
Specifications
- 容量:5 ml
- 顺序:04
- 分度:0,1 ml
- 误差:0,045 ± ml