订单输入
China
Orders LinkContactUsLinkComponent
Dissection boards

Dissection boards

02, Dissection board, 白色, 610×460×25 mm
目录编号 BOCH12616
供应商:   BOCHEM
Dissection boards
目录编号 BOCH12616
供应商:  BOCHEM
Dissection boards
Dissection boards

02, Dissection board, 白色, 610×460×25 mm

Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents

Hygienic dissection boards made from PE.

  • Two colours available
  • Suitable for dish washer - Heat resistant up to 90 °C

HACCP compliant

 : HACCP compliant

规格
  • 颜色:
    白色
  • 说明:
    Dissection board
  • 顺序:
    02
  • 宽度×深度×高度:
    610×460×25 mm